Magnetic Molecular Model Set, 36 Atoms
470231-446
- Description:Magnetic Molecular Model Set, 36 Atoms
- Cat. no.:470231-446
Perfect for Magnetic Boards for Classroom Demonstrations
- 36 atoms
- 10 magnets
This set of 10 magnetic molecules includes all molecular models for studying combustion reactions, methane and butane. The set comes with 10 powerful magnets. To insert the magnets please follow procedure outlined in enclosed instructions.
Contents: 2 Black 0 Face Magnetic Carbon (38 mm), 2 Black 2 Face Magnetic Carbon (38 mm), 1 Black 3 Face Magnetic Carbon (38 mm), 1 Black 3 Face Magnetic Carbon (38 mm), 2 Red 1 Face Magnetic Oxygen (38 mm), 2 Red 2 Face Magnetic Oxygen (38 mm), 17 White O Face Non-Magnetic Hydrogen (30 mm), 6 Red 0 Face Non-Magnetic Oxygen (38 mm), 3 Black 3 Face Non-Magnetic Carbon (38 mm), 10 Magnets.