SMT Stencil Wipe, Premium Grade, Large, MicroCare
Catalog # 76020-252
Supplier: MICROCARE CORP.
Specifications
- Description:SMT Stencil Wipe, Premium Grade, Large
- Cat. no.:76020-252
Specifications
About this item
This is the premier, highest-quality general purpose stencil wipe in the industry.
- SMT stencil wipe, highest quality, best scrubbing, least lint, large size
- Very strong, no cellulose, no polyster and no glues, so less linting and better cleaning
- Cleaner, stronger, more absorption than competing wipes
- High absorption, medium weight, excellent low-lint material, excellent purity
Featuring the exclusive MicroWipe™ FP fabric, the wipe has no polyester, cellulose or glues, which means it delivers much better cleaning with less lint. Approximately 50% more absorption and 80% less linting than the benchmark wipes.
The larger wipe (but not the largest) sized for cleaning SMT stencils and other chores involving large, flat surfaces. It’s the optimal size for use on stencils and other maintenance chores like cleaning pallets and reflow ovens, but it’s probably too big to use on most modern PCBs. Class 1000 clean room packaging. Large, money-saving package.
Packaging: 300 sheets/bag, 12 bags/case.