Order Entry
Mexico
Orders LinkContactUsLinkComponent
General Purpose Wipe, Electronics Grade, Medium Size, MicroCare
General Purpose Wipe, Electronics Grade, Medium Size, MicroCare
General Purpose Wipe, Electronics Grade, Medium Size, White, 50 Sheets/Bag
Catalog # 76020-239
Supplier:   MICROCARE CORP.
Supplier #:  MCC-W66
General Purpose Wipe, Electronics Grade, Medium Size, MicroCare
Catalog # 76020-239
Supplier:  MICROCARE CORP.
General Purpose Wipe, Electronics Grade, Medium Size, MicroCare
General Purpose Wipe, Electronics Grade, Medium Size, MicroCare
General Purpose Wipe, Electronics Grade, Medium Size, White, 50 Sheets/Bag
Price
$589.45
Log in for availability
Paquete de 55 Case de 25 (55UN)
Product Details & Documents
This is an excellent benchtop wipe sized for the rework bench and a perennial MicroCare best-seller.
  • Polyester and cellulose construction
  • 6×6" (150×150 mm)
  • Medium (2.1 oz.) weight
  • Economically priced, convenient package size
The 76020-241 part number is the exact same wipe in a jumbo-sized, economy package. Save about 30% per wipe with the economy packaging.
Medium absorption, medium weight, relatively lint-free, good purity. While it’s the optimal size for use on the rework bench, it’s probably too small for stencil cleaning. Smooth, white, medium absorbency, standard packaging.
Specifications
  • Color:
    White
  • Description:
    General Purpose Wipe, Electronics Grade, Medium Size
  • Packaging:
    50 Sheets/Bag