Difco™ M17 Agar, BD Diagnostics
M 17 Agar, 500 g
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Supplier #: 218571
Difco™ M17 Agar, BD Diagnostics
Catalog # 95056-558
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Jump to:
Product Details & Documents
Use for the cultivation and isolation of mesophilic lactic streptococci and for the selection of Streptococcus thermophilus from yogurt, cheese starters and other dairy products.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:M 17 Agar
- Size:500 g