Order Entry
Mexico
Orders LinkContactUsLinkComponent
Protein Test Strips
Protein Test Strips
Protein Test Strips, 50
Catalog # 470092-374
Número de proveedor #:  PRO-1V-50
Protein Test Strips
Catalog # 470092-374
Protein Test Strips
Protein Test Strips
Protein Test Strips, 50
Price
$361.92
Log in for availability
Paquete de 1
Product Details & Documents
Add protein testing to your urinalysis procedures.
  • Flip top vial
  • Waterproof color chart
These single-test test strips have a plastic strip with a reactive pad to prevent contact contamination while handling. Simply dip the pad into the solution, wait 60 seconds, and compare to the color chart on the side of the bottle.
Specifications
  • Description:
    Protein Test Strips
  • No. of Tests:
    50