Disodium hydrogen phosphate dihydrate ≥98.5% for HPLC, Fluka™
Catalog # BJ71633-250G
Supplier: HONEYWELL INTERNATIONAL INC MX
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pack type:Plastic bottle
- Size:250 g
- Cat. no.:BJ71633-250G
- Supplier No.:71633-250G
Specifications
Specification Test Results
- Assay:98.5 - 102.0 %
- pH (10 %):8.8 - 9.4
- Colour of the solution:complying
- Turbidity of solution:complying
- Absorbance 500 nm:Max. 0.02
- Absorbance at 320 nm:Max. 0.02
- Absorbance 280 nm:Max. 0.04
- Absorbance at 260 nm:Max. 0.06
- Absorbance at 230 nm:Max. 0.1
Specification Test Results
About this item
:
:177.99 g/mol
Melting Pt:450 °C
Density:2.07 g/cm³ (16 °C)
Storage Temperature:Ambient
CAS Number:10028-24-7
Formula:Na₂HPO₄·2H₂O
MDL Number:MFCD00149182