Order Entry
Mexico
Orders LinkContactUsLinkComponent
Hooke’s Law Spring Set

Hooke’s Law Spring Set

Hooke’s Law Spring Set
Catalog # 470230-122
Supplier #:  PH0323SS
Hooke’s Law Spring Set
Catalog # 470230-122
Hooke’s Law Spring Set
Hooke’s Law Spring Set

Hooke’s Law Spring Set

Price
$189.77
Log in for availability
Each (1UN)
Product Details & Documents

Hooke's Law spring set with variety of spring constants.

  • Set of 5 springs, each with a different spring constant
  • Great for inquiry lab investigations
  • Length 5 cm
  • Inner diameter 8 mm

This set of five springs is great for in-depth studying of the principles of Hooke's Law. Each spring has a different constant. For use with Labs Hooke's Law Apparatus.

Individual springs:
10 N/m (max 100 g);
20 N/m (max 225 g);
50 N/m (max 550 g);
100 N/m (max 1100 g);
200 N/m (max 2200 g).

Specifications
  • Description:
    Hooke’s Law Spring Set