Belts for Wimshurst
Belts for Wimshurst, 2 pk.
Catalog # 470230-304
Supplier: EISCO SCIENTIFIC LLC
Supplier #: PH0848BELT
Jump to:
Product Details & Documents
Spare belt for Wimshurst machine. This item has two replacement belts, both of which have a 20" circumference.
- Smaller dia. belt dia. 0.13"
- Larger dia. belt dia. 0.20"
- Black in color
- Flexible rubber material
The flexible rubber belts is for use with the Wimshurst Machine (SKU# PH0848C or SKU# PH0848D) - this machine (with the belts) is a hand-cranked electrostatic generator that produces voltage and, in essence, creates lightening!
Specifications
- Description:Belts for Wimshurst, 2 pk.



