Order Entry
Mexico
Orders LinkContactUsLinkComponent
Plotting Compass, 100 mm

Plotting Compass, 100 mm

Plotting Compass, 100 mm
Catalog # 470230-288
Supplier #:  PH0820F
Plotting Compass, 100 mm
Catalog # 470230-288
Plotting Compass, 100 mm
Plotting Compass, 100 mm

Plotting Compass, 100 mm

Price
$120.99
Log in for availability
Each (1UN)
Product Details & Documents

Aluminum, economical plotting compass.

  • 4 inch (100 mm) aluminum plotting compass with glass face
  • Economically priced
  • Great option for students
  • Dial marked with principle points

This 4 inch aluminum plotting compass is marked with principle points and has a glass face. It is economically priced and great for classroom use.

Specifications
  • Description:
    Plotting Compass, 100 mm