BD Trypticase™ Soy Agar, BD Diagnostics
90005-170
:
- Description:BD Trypticase™ Soy Agar
- Size:500 mL
- Cat. no.:90005-170
- Type:Screw Cap
- Supplier No.:299099
BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples
- Use of highest quality BBL and Difco brand dehydrated culture media for preparation
- Conformation to current USP and EP requirements (see package labeling for details)
- Color-coded caps and/or labeling for most products, providing easy product identification
- Transparent labeling to allow a clear view of media and culture
Screw Cap bottle has a black, phenolic screw cap.
They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.