Order Entry
Mexico
ContactUsLinkComponent
Fluid D Prepared Bottled Media, BD Diagnostics
Fluid D Prepared Bottled Media, BD Diagnostics
Catalog # 90004-130
CAS Number:  
undefined
Fluid D Prepared Bottled Media, BD Diagnostics
Catalog # 90004-130
Supplier Number:  290662
CAS Number:  

Specifications

  • Description:
    Fluid D, 0.1% Peptic digest of animal tissue and Polysorbate 80
  • Size:
    300 mL
  • Cat. no.:
    90004-130
  • Type:
    Septum/Screw Cap
  • Supplier No.:
    290662

Specifications

About this item

BD Prepared Bottled Media are ready-to-use formulations intended for the detection of micororganisms from pharmaceutical, food, environmental or water testing samples. They are offered in a wide variety of bottle styles and fill volumes designed to meet customer requirements.

  • Use of highest quality BBL and Difco brand dehydrated culture media for preparation
  • Conformation to current USP and EP requirements (see package labeling for details)
  • Color-coded caps and/or labeling for most products, providing easy product identification
  • Transparent labeling to allow a clear view of media and culture

Serum bottle holds up to 100 mL fill volume.

Septum/Screw Cap style has a black, phenolic screw cap with septum.