Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cleaning Agents
Detergents and Surface Cleansers
Vertrel® XF Solvent, MicroCare
Vertrel® XF Solvent, MicroCare
46620-714
:
MICROCARE CORP.
Small Business Enterprise
Vertrel® XF Solvent, MicroCare
46620-714
:
MICROCARE CORP.
:
MCC-XFG
Small Business Enterprise
Description:
Vertrel® XF Solvents for Vapor Cleaner
Cat. no.:
46620-714
Weight:
4.54 kg
Supplier No.:
MCC-XFG
Nonflammable
Plastic-safe
Ozone safe for ultrasonic tanks