Order Entry
Mexico
ContactUsLinkComponent
Tissue-Tek® Uni-Cassette® Cassette Standard Systems, Sakura® Finetek
Tissue-Tek® Uni-Cassette® Cassette Standard Systems, Sakura® Finetek
Catalog # 14221-384
Tissue-Tek® Uni-Cassette® Cassette Standard Systems, Sakura® Finetek
Catalog # 14221-384
Supplier Number:  4170-01

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Color:
    White
  • Cat. no.:
    14221-384
  • Supplier No.:
    4170-01

Specifications

About this item

The Tissue-Tek® Uni-Cassette® standard cassette system is utilized to provide specimen identification and security from the grossing station to the paraffin block. In these systmes smooth, single-action fracture-back hinges are present for greater protection. The LWS (Large Writing Surface) Uni-Cassette® are used when a larger writing front surface is needed.

  • Designed for maximum fluid exchange during processing
  • Contains a base and attached lid, which is easily removed
  • Manufactured from chemical-resistant plastic including solvents and decalcifying solutions
  • Raised spacer nubs for better exchange of processing reagents
  • Wider finger tabs to reduce strain

Reusable stainless steel base molds, for use with Uni-Cassette® and Uni-Cassette® Biopsy, are 5 mm deep, and feature extended fins to prevent tipping and to provide a gripping surface

Uni-Cassettes can be printed on the Tissue-Tek® SmartWrite® and Tissue-Tek® AutoWrite® Printers. Large writing surface cassettes are not compatible with SmartWrite® Cassette Printer.

Flat metal bottom allows maximum heat/cold exchange.