Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Especificaciones
- Description:Hydroponics Test Kit
- Cat. no.:470228-880
- Range:pH 4.5 - 8.0,
Nitrate Nitrogen 5 - 200 ppm,
Phosphorous 3 - 30 ppm,
Sulfate 0 - 200 ppm,
Potassium 0 - 250 ppm - No. of Tests:50
Especificaciones
Acerca de este artículo
The Hydroponics 4-Way Test Kit offers tests for pH and three key nutrient factors: nitrogen, phosphorus and potassium.
- Includes materials to perform 50 tests
- Offers tests for pH, nitrogen, phosphorus, and potassium
- 75 page handbook included
It allows the hydroponic hobbyist to maintain proper nutrient balance and to achieve optimum growing conditions in soil-less cultures. Reagents sufficient for 50 tests per factor and complete labware are foam-mounted in a sturdy carrying case. Instructions and 75-page hydroponics handbook are supplied.