Single Element Standard, Zinc for AA and ICP-OES, Pure Grade, PerkinElmer
97012-020
:
- Concentration:1000 µg/ml
- Matrix:2% HNO₃
- Size:125 ml
- Environmentally Preferable:
- Cat. no.:97012-020
- Element:Zinc
- Supplier No.:N9300178
- Analyzed by ICP-OES
- Analyzed by Classical Wet Assay
- 32 trace impurities analyzed by ICP-MS of the final solution and reported on the certificate
- Impurities reported at ppm level
- All Standards are prepared and certified under ISO Guide 34 and ISO 17025 – certified by A2LA