Order Entry
Mexico
Orders LinkContactUsLinkComponent
Balance Forceps, Walter Stern

Balance Forceps, Walter Stern

Curved, 10 cm (4")
Catalog # 25672-100
Supplier:   WALTER STERN INC
Supplier #:  300-006
Balance Forceps, Walter Stern
Catalog # 25672-100
Supplier:  WALTER STERN INC
Balance Forceps, Walter Stern

Curved, 10 cm (4")

Price
$93.47
Log in for availability
Each (1UN)
Product Details & Documents

Stainless steel curved forceps with plastic tips.

Specifications
  • Description:
    Curved
  • Length:
    10 cm (4")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.