Order Entry
Mexico
ContactUsLinkComponent
Fendall® Flash Flood® Emergency Eyewash Stations, Honeywell Safety
Fendall® Flash Flood® Emergency Eyewash Stations, Honeywell Safety
Catalog # 56611-100
undefined
Fendall® Flash Flood® Emergency Eyewash Stations, Honeywell Safety
Catalog # 56611-100
Supplier Number:  32-000400-0000

Specifications

  • Width:
    14"
  • Capacity:
    1 gal.
  • Description:
    Eyewash Station
  • Height:
    20"
  • Length:
    10½"
  • Cat. no.:
    56611-100
  • Supplier No.:
    32-000400-0000
  • Flow:
    3 min

Specifications

About this item

The Fendall Flash Flood delivery system offers immediate, hands-free eye flushing, and is portable to meet the needs of changing work-place hazards.

  • One-gallon sealed cartridges are easy to store
  • Easy installation with no plumbing required
  • Dual nozzles provide simultaneous flushing to both eyes for three minutes
  • Highly portable system may be easily wall mounted

Unique cartridge design delivers a buffered, saline solution superior to tap water for emergency eye care. Nozzle strap is easily removed for hands-free flushing, leaving hands free to hold eyelids open. Eyesaline solution cartridges, once installed, require a 6-month maintenance cycle. Unopened cartridges have a shelf life of up to 24 months.

Can be easily wall mounted for placement near most workplace hazards.