Order Entry
Mexico
ContactUsLinkComponent
Difco™ Salmonella (Somatic) Antisera O, Lyophilized, BD Diagnostics
Difco™ Salmonella (Somatic) Antisera O, Lyophilized, BD Diagnostics
Catalog # 90002-054
CAS Number:  
Difco™ Salmonella (Somatic) Antisera O, Lyophilized, BD Diagnostics
Catalog # 90002-054
Supplier Number:  230191
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Description:
    Salmonella O Antiserum, Factors 1, 3, 19, Group E4
  • Size:
    3 ml
  • Cat. no.:
    90002-054
  • Supplier No.:
    230191

Specifications

About this item

Difco™ Salmonella O Antisera are lyophilised, polyclonal rabbit antisera containing approximately 0.04% Thimerosal as a preservative

  • Store at 2 to 8°C (35.6 to 46.4°F)
  • Contains sufficient reagents for 60 tests

Difco™ Salmonella O Poly Antisera are polyvalent antisera. Each antiserum is specific for certain serogroup antigens.