Fractional Weight Set
1×1 mg, 2×2 mg, 1×5 mg, 1×10 mg, 2×20 mg, 1×50 mg, 1×100 mg, 2×200 mg, 1×500 mg
Catalog # 470144-652
Supplier: UNITED SCIENTIFIC SUPPLIES
Número de proveedor #: FWB500-S
Jump to:
Product Details & Documents
Milligram Weights for your Classroom.
- Plastic storage case included
This fractional weight set is composed of 12 different items ranging in mass from 1mg to 500mg.
Specifications
- No. of Weights:1×1 mg, 2×2 mg, 1×5 mg, 1×10 mg, 2×20 mg, 1×50 mg, 1×100 mg, 2×200 mg, 1×500 mg