Order Entry
Mexico
ContactUsLinkComponent
Heavy Support Stands with Rods, Stamped Steel
Heavy Support Stands with Rods, Stamped Steel
Catalog # 470144-202
undefined
Heavy Support Stands with Rods, Stamped Steel
Catalog # 470144-202
Supplier Number:  SSB4X6-H

Specifications

  • Rod Length:
    46 cm
  • Description:
    Support Stand with Rod
  • Cat. no.:
    470144-202
  • Base Dimensions:
    10×15 cm
  • Supplier No.:
    SSB4X6-H

Specifications

About this item

Sturdy and heavy support stands.

  • Extra heavy base
  • Thicker screw hole mounts
  • Includes a support rod

Heavy stamped steel bases with black enamel finish feature an added steel plate on the back, which makes them about 50% heavier than regular stamped steel bases. The steel plate also facilitates a sturdier threading of the rod into the base.

Delivery: Includes a removable, zinc-plated steel rod.