Order Entry
Mexico
ContactUsLinkComponent
DNA/RNA Shield™ Reagent, Zymo Research
DNA/RNA Shield™ Reagent, Zymo Research
Catalog # 76020-418
Supplier:  Zymo Research
CAS Number:  
undefined
DNA/RNA Shield™ Reagent, Zymo Research
Catalog # 76020-418
Supplier:  Zymo Research
Supplier Number:  R1100-250
CAS Number:  

Specifications

  • Method / Format:
    Chemical extraction & preservation
  • Prep size:
    Scalable
  • Refine target molecule:
    Total RNA
    Total DNA
  • Sample type:
    Stool
    Swabs
    Biofluids (serum, plasma, saliva)
    Cultured cells
    Human/animal cells and tissues
  • Target molecule:
    Copurification of DNA & RNA
    DNA
    RNA
  • Description:
    DNA/RNA Shield™ (1X Concentrate)
  • Size:
    250 ml
  • Description:
    DNA/RNA Shield™ (1X Concentrate)
  • Cat. no.:
    76020-418

Specifications

About this item

DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures.

  • Nucleic acid preservation (at ambient temperature; cold-free)
  • Pathogen inactivation (bacteria, fungus, parasites & viruses)
  • RNA Stability: Ambient temperature (4 to 25 °C) >1 month
  • DNA Stability: Ambient temperature (4 to 25 °C) >2 years
  • DNA and RNA: Frozen (<−20 °C): Indefinitely

DNA/RNA Shield™ ensures nucleic acid stability during sample storage/transport at ambient temperatures. There is no need for refrigeration or specialized equipment. DNA/RNA Shield™ effectively lyses cells and inactivates nucleases and infectious agents (virus), and it is compatible with various collection and storage devices (vacutainers, swabs, nasal, buccal, fecal, etc.).