Order Entry
Mexico
ContactUsLinkComponent
Ammonium Test, MColortest, MilliporeSigma
Ammonium Test, MColortest, MilliporeSigma
Catalog # EM1.14428.0002
CAS Number:  
Ammonium Test, MColortest, MilliporeSigma
Catalog # EM1.14428.0002
Supplier Number:  1144280002
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Indophenol blue
  • Description:
    Ammonium Test
  • Storage temperature:
    Store at +15°C to +25°C.
  • Cat. no.:
    EM1.14428.0002
  • Application:
    Ammonium Ions and Dissolved Ammonia in Ground Water and Surface Water;Seawater;Drinking Water and Aquarium Water
  • Range:
    0,025-0,05-0,075-0,1-0,15-0,2-0,25-0,3-0,4 mg/l NH₄
  • Supplier No.:
    1.14428.0002
  • No. of Tests:
    70

Specifications

About this item

MColortest™ with colour card comparator (formerly Aquaquant®).

By using cuvettes with a long light path, the MColortest™ test kits are especially suited for measuring low ion concentrations (some as low as 0,005 mg/l). These test kits are especially appropriate for analysing drinking water, spring water, purified and mineral water, as well as cooling and boiler water.