Order Entry
Mexico
ContactUsLinkComponent
PCR Tubes, Watson Bio Lab
PCR Tubes, Watson Bio Lab
  75982-954
undefined
PCR Tubes, Watson Bio Lab
  75982-954
 :  137-212C
  • Cap Type:
    Dome-shaped cap
  • Capacity:
    0.2 ml
  • Color:
    Natural
  • Description:
    Individual PCR tubes
  • Cat. no.:
    75982-954
The side walls are uniformly thin, which enables PCR with superior heat exchange.
  • Thin wall, high transparency
  • Nuclease-free certified
Flat and dome caps are available.
All tubes feature autoclavable polypropylene construction.
The flat top, smooth and superior in optical transparency, is the most suitable for Real Time PCR. They also offer writing space.