Order Entry
Mexico
Orders LinkContactUsLinkComponent
Round-Bottom Standard Microtubes, Watson Bio Lab
Round-Bottom Standard Microtubes, Watson Bio Lab
Round-Bottom Standard Microtube, 0.6 ml
Catalog # 75982-888
Supplier:   WATSON BIO LAB USA
Supplier #:  130-806C
Round-Bottom Standard Microtubes, Watson Bio Lab
Catalog # 75982-888
Supplier:  WATSON BIO LAB USA
Round-Bottom Standard Microtubes, Watson Bio Lab
Round-Bottom Standard Microtubes, Watson Bio Lab
Round-Bottom Standard Microtube, 0.6 ml
Price
$926.83
Log in for availability
Paquete de 1,000 Case de 10 (1,000,UN)
Product Details & Documents
These user-friendly standard microtubes feature an easy-opening, flat-top cap.
  • Flat Cap
  • High Transparency
  • RNase-/DNase-/Human DNA-free
  • Autoclavable Polypropylene Construction
  • Scale Graduations
The flat cap provides a space to write. Users can autoclave the polypropylene tube. Graduated scale makes measuring simple.
Accessories information: Pestles, storage racks, and cap locks are available separately.
Specifications
  • Volume:
    0.6 ml
  • Description:
    Round-Bottom Standard Microtube