Order Entry
Mexico
Orders LinkContactUsLinkComponent
Linear Diffraction Glasses
Linear Diffraction Glasses
Diffraction Glasses, Linear, 500 lines/mm
  470225-846
Supplier #:  01616
Linear Diffraction Glasses
  470225-846
Linear Diffraction Glasses
Linear Diffraction Glasses
Diffraction Glasses, Linear, 500 lines/mm
$186.54
Paquete de 10
Make the study of light and color fun and easy
  • Uses a linear diffraction grating
  • Easily see identifiable spectral images
Highly efficient, 500 lines per millimeter, single axis holographic diffraction grating lenses separate light from any source into its spectral components for study and analysis. Recommended for use with Power Supply and Spectrum Gas Tubes.
  • Description:
    Diffraction Glasses, Linear, 500 lines/mm