Dissolution Paddles, Apparatus 2, Quality Lab Accessories
11010-396
- Description:19’’ (480 mm) Paddle for VanKel/Varian, Serialized
- Material:PTFE coated over 316 Stainless Steel
- Cat. no.:11010-396
- Supplier No.:NPD019-01
QLA offers a variety of Paddles for most dissolution baths.
- Made of high quality 316 electropolished stainless steel, PTFE coated over 316 SS or solid fluoropolymer
- All paddles made in the USA
- All paddles are laser marked with a unique serial number
- Other paddle shaft lengths are available
All styles comply with USP (711) and Original Equipment Manufacturer's dimensional specifications. The single piece molded Fluoropolymer paddles provide a seamless construction eliminating corrosion and contamination encountered with conventional three-piece assemblies. All QLA paddles are precision manufactured to limit rotational wobble to levels below USP (711) specifications.
: All paddles are supplied with a Certificate of Conformance (COC) stating that they are USP compliant and that they meet machine manufacturer specifications. A Certificate of Analysis (COA) is available at an additional cost.