Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Media for Microbiology
Dehydrated Media for Microbiology
OmniPur® Casamino Acids, MilliporeSigma
OmniPur® Casamino Acids, MilliporeSigma
OmniPur® Casamino Acids, MilliporeSigma
Catalog # EM-2240
Supplier:
THOMAS SCIENTIFIC INC MX
CAS Number:
Certificates
Expand image
OmniPur® Casamino Acids, MilliporeSigma
Catalog # EM-2240
Supplier:
THOMAS SCIENTIFIC INC MX
Supplier Number:
2240-500GM
CAS Number:
Certificates
Specifications
Pack type:
Plastic bottle
Size:
500 g
Cat. no.:
EM-2240
Supplier No.:
2240-500GM
Specifications
About this item
Product Details
Documentation
Product Details
Documentation