Dipping Bottle, Bürkle
Dipping Bottle, 1000 ml (33.81 oz.), 148 mm (5.83")
Catalog # 75846-222
Supplier: BURKLE INC
Supplier #: 5365-4000
Jump to:
Product Details & Documents
Dipping bottle with glass container for sampling in sewage plants, as well as mud and water samples.
- Equipped with Easy-Click; a safe and practical closure for a rapid bottle exchange
- High weight (3.3 kg/7.28 lbs.) for fast lowering
- Height with handle is 330 mm (12.99")
- Tested for use in the Ex Protection Zone 0
For testing crude oil, liquid mineral oil products, fuels, special and test fuels, fuel oils, petroleum, and liquid lubricants of Hazard Categories IIA and IIB. Constructed from completely non-sparking materials. Dipping frame made of nickel-plated brass, with an easy-click closure made of electrically conductive polyamide.
Packaging: Supplied one dipping bottle per package.
Documents
Dipping Bottle
Videos
Specifications
- Ø:148 mm (5.83")
- Capacity:1000 ml (33.81 oz.)
- Description:Dipping Bottle
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



