Magnetizer Box
Magnetizer Box
Catalog # 470159-586
Supplier: EISCO SCIENTIFIC LLC
Supplier #: MAGBARV1
Jump to:
Product Details & Documents
No need to Buy New Magnets
- Bar Magnet
- 110/120V AC
- 60Hz
Revitalizes iron, nickel, cobalt, and alnico horseshoe and bar magnets in seconds. Simply place magnets in the 25x25 mm holes and press the button to make old magnets new again.
Specifications
- Description:Magnetizer Box