Energy Stick
Energy Stick, 7 x 1.5"
Catalog # 470223-984
Supplier: CREATIVE KIDS FAR EAST INC.
Supplier #: 7250
Jump to:
Product Details & Documents
Energize your lessons on electricity.
- Demonstrate open and closed circuits
- No danger of shock
- Colored lights
Safely demonstrate open and closed series circuits without any danger of electric shocks! When the tube is 'turned on,' the red, green, and blue lights light up and it makes a funny buzzing sound. Activating the tube is the fun part - just touch the metal ends with both hands. Your body forms the conducting material that bridges the gap between the two metal strips and closes the circuit!
Specifications
- Description:Energy Stick
- Size:7 x 1.5"