Order Entry
Mexico
Orders LinkContactUsLinkComponent
Flying Pig
Flying Pig
Flying Pig
Catalog # 470223-114
Supplier:   ARBOR SCIENTIFIC
Supplier #:  P4-2165
Flying Pig
Catalog # 470223-114
Supplier:  ARBOR SCIENTIFIC
Flying Pig
Flying Pig
Flying Pig
Price
$669.46
Log in for availability
Each (1UN)
Product Details & Documents
Make students learn when pigs fly.
  • Includes mounting string
  • Includes instruction manual
Add some excitement to your lessons on circular motion with the Flying Pig. Suspended from the ceiling, his wings flap and carry him in a regular circle. With the string, this motion gives students a conical pendulum and a perfect opportunity for quantitative measurements of circular motion.
Ordering information: Lab instructions included.
Specifications
  • Description:
    Flying Pig