Difco™ D-Mannitol, BD Diagnostics
Catalog # 90000-934
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Specifications
- Description:D-Mannitol
- Size:500 g
- Cat. no.:90000-934
- Supplier No.:217020
Specifications
About this item
D-Mannitol is an osmotic diuretic agent that shows diuretic effects in vivo.
- Orally active
- Color: White
D-Mannitol inhibits renal tubular reabsorption of sodium, chloride, and other solutes. It also inhibits neutrophil apoptosis.