Order Entry
Mexico
ContactUsLinkComponent
Acetonitrile ≥99.9% CHROMASOLV™, gradient grade for HPLC
 
 
Acetonitrile ≥99.9% CHROMASOLV™, gradient grade for HPLC
  BJ34851-4L
   Safer Human and Environmental Health
 :  
undefined
 
 
Acetonitrile ≥99.9% CHROMASOLV™, gradient grade for HPLC
  BJ34851-4L
 :  34851-4X4L
   Safer Human and Environmental Health
 :  

 

  • Pack type:
    Glass bottle
  • Size:
    4x 4 L
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 solvent selection guide recommended by default
  • Cat. no.:
    BJ34851-4L
  • Supplier No.:
    34851-4X4L

 

 

  • Assay (GC):
    Min. 99.90 %
  • Refractive index (n 20/D):
    1.3435 - 1.3445
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.02 %
  • Free acid (as CH₃COOH):
    Max. 0.001 %
  • Free alkali (as NH₃):
    Max. 0.0002 %
  • Absorbance at 400 nm:
    Max. 0.0044
  • Absorbance at 250 nm:
    Max. 0.0044
  • Absorbance at 235 nm:
    Max. 0.0044
  • Absorbance at 230 nm:
    Max. 0.0044
  • Absorbance at 200 nm:
    Max. 0.032
  • Absorbance at 195 nm:
    Max. 0.12
  • Fluorescence (chinin) at 254 nm:
    Max. 1 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 0.5 ppb
  • HPLC Gradient at 210 nm:
    Max. 3 mAU
  • HPLC Gradient at 254 nm:
    Max. 0.5 mAU
  • Baseline drift at 210 nm:
    Max. 15 mAU
  • Refractive Index:
    n20/D 1.344 (lit.)
  • Measuring Range:
    ≤15 mAU baseline drift, (210 nm)
  • Autoignition Temperature:
    973 °F

 

 

 :75-05-8
 :
 :H₃CCN
 :MFCD00001878
 :41.05 g/mol
 :82 °C (1.013 hPa)
 :81-82 °C (lit.)
 :–46 °C
 :−48 °C (lit.)
 :0.786 g/mL at 25 °C (lit.)
 :6 °C
 :2 °C
 :Ambient
 :200-835-2
 :1648
 :3,II
 :GHS02
 :GHS07