BioClean™ MTA™ Sterile Tie-On Facemask, Ansell
Catalog # 75836-210
Supplier: Ansell Healthcare
Specifications
- Sterility:Sterile
- Material:PE
- Description:BioClean MTA™ sterile facemask with ties
- Cat. no.:75836-210
- Supplier No.:MTA210-1
Specifications
About this item
BioClean MTA™ Facemask is latex-free and low-linting.
- Ultra-low particulation
- High strength seams
- 3-ply design with ultrasonic sealing
- High BFE and PFE
- Outer Facing Layer: Non-woven spunbonded PE (hydrophobic)
Providing high particle and bacterial filtration efficiency, the latex-free and low-linting BioClean MTA™ Sterile Facemask with Ties combines reliable protection with comfort and exceptional breathability.
Certifications: ISO Class 4 and EU GMP Grade A compatible.
Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Sampling and inspection in accordance with ISO 2859-3:1994.
Tested against standard ASTM F2101 for Bacterial Filtration Efficiency (BFE).
Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Sampling and inspection in accordance with ISO 2859-3:1994.
Tested against standard ASTM F2101 for Bacterial Filtration Efficiency (BFE).
Packaging: Face masks are packaged one piece per sealed inner PE bag; 50 inner bags per sealed outer PE bag; 10 outer bags per lined carton (500 pieces).