About this item
Take simple conductivity, TDS and temperature measurements with the Orion™ Star™ A112 Conductivity Benchtop Meter, which offers easy operation for basic laboratory testing.
- AUTO-READ™ technology locks in the stable reading on screen and a ready indicator alerts users when readings are stable
- Included electrode arm and newly designed holder make it easier to maintain and place probes into samples
- Meter plugs into almost any power outlet with the included universal power adapter, or use four AA batteries
- Nonvolatile memory holds up to 50 data points
- IP54-rated housing withstands splashes
- Meter mounts to wall, saving bench space
- Three-year meter warranty
Take simple, routine conductivity, total dissolved solids (TDS), and temperature measurements with the Orion™ Star™ A112 Conductivity Benchtop Meter, an easy-to-use and budget-friendly instrument for basic conductivity analysis in the laboratory.
The A112 Conductivity Benchtop Meter features a selectable cell constant as well as selectable reading reference temperatures of 20 or 25°C to ensure accurate results. The large, informative LCD screen clearly shows the main information, including conductivity or TDS reading, temperature, meter mode, and power source.
Obtain temperature compensated readings using linear option with 20 or 25°C reference temperatures. Quickly navigate setup menus and ensure consistent calibrations using the simple keypad layout and on-screen text prompts. Use the included electrode stand to easily place sensors in samples and simplify storage.