Order Entry
Mexico
ContactUsLinkComponent
 
Aldrin, Restek
  75834-236
 :  
Aldrin, Restek
  75834-236
 :  32205
 :  

 

  • Concentration:
    1000 µg/ml in methanol
  • CAS No.:
    309-00-2
  • Description:
    Aldrin
  • Size:
    1 ml/Ampule
  • Formula:
    C₁₂H₈Cl₆
  • Description:
    Aldrin
  • Cat. no.:
    75834-236
  • Supplier No.:
    32205

 

 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.