Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Ethylene Oxide, Restek

Ethylene oxide, 50 mg/ml in methylene chloride, 1 ml/Ampule
  75834-514
 :  30620
Ethylene Oxide, Restek
  75834-514
 :  
Ethylene Oxide, Restek

Ethylene oxide, 50 mg/ml in methylene chloride, 1 ml/Ampule

 :  
 
$3,389.80
   
Each (1UN)
 
   
 

The test for ethylene oxide is specified in many individual drug monographs of USP24/NF19. The limit test concentration is currently 10 ppm. While the specific test solution and method will vary depending on the particular drug monograph, the solution below is suitable for most tests.

 : Ethylene oxide is available in other solvents and concentrations. Request your custom formulation at www.restek.com/solutions

 

 

 

  • Concentration:
    50 mg/ml in methylene chloride
  • Description:
    Ethylene oxide
  • Size:
    1 ml/Ampule
  • Cat. No.:
    75834-514
  • Formula:
    C₂H₄O
  • Description:
    Ethylene oxide
  • CAS No.:
    75-21-8

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.