New designed high wattage, constant temperature VWR® standard dry block heaters with smaller footprint are economical, versatile and compact.
Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures
120 V model
Exceptional temperature uniformity
Holds interchangeable modular heating blocks
Heats to 150 °C (302 °F)
5-Year-Warranty on parts & labor
Manufactured in the USA
Each model accepts separate interchangeable modular blocks, which accommodate various sample enclosures such as vials, microplates, PCR strips, and a wide variety of tubes. Each block contains a thermometer well, and features close tube-and-block contact for maximum heat retention.
All units offer two separate temperature adjustment knobs providing reproducible low temperature and high temperature control. Low range knob adjusts from ambient to 100 °C (212 °F) and high range knob adjusts from 75 to 150 °C (167 to 302 °F).
These high-wattage, constant-temperature block heaters are economical and versatile. Each unit requires minimum bench space and features convenient front-mounted temperature adjustment knobs providing high and low temperature control.
- Catalog No:
- 75838-318
- 75838-286
- 75838-298
- Environmentally Preferable:
- Sustainable Packaging
- Waste Reducing
- Waste Reducing
- Temperature range:
- 5° above ambient to 150°C (302°F)
- 5° above ambient to 150°C (302°F)
- 5° above ambient to 150°C (302°F)
- Temperature stability:
- ±1°C (37 °C)
- ±1°C (37 °C)
- ±1°C (37 °C)
- Temperature uniformity:
- ±0.1°C (within the Block @ 37°C)
- ±0.1°C (within the Block @ 37°C)
- ±0.1°C (within the Block @ 37°C)
- Environmental certification:
- RoHS
- RoHS
- RoHS