Order Entry
Mexico
ContactUsLinkComponent
Enterobacterial Enrichment (EE) Broth Mossel, BD Life Sciences
Enterobacterial Enrichment (EE) Broth Mossel, BD Life Sciences
Catalog # 90003-524
CAS Number:  
Enterobacterial Enrichment (EE) Broth Mossel, BD Life Sciences
Catalog # 90003-524
Supplier Number:  256620
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EE Broth Mossel
  • Size:
    500 g
  • Cat. no.:
    90003-524
  • Supplier No.:
    256620
Specifications
About this item
EE Broth Mossel is used for selective enrichment of Enterobacteriaceae in bacteriological examination of foods.
  • Peptones provide nitrogen, vitamins, and amino acids
  • Dextrose is a carbon source
  • Disodium phosphate and monopotassium phosphate are buffering agents
  • Brilliant green and oxgall are selective agents
EE Broth Mossel is listed in the USP as one of the recommended media for the isolatoin of bile-tolerant gram-negative bacteria from nonsterile pharmaceutical products.