Specifications
- Description:Frameless glasses
- Cat. no.:89135-117
- Lens Coating:Anti-fog
- Lens Tint:Clear
- Supplier No.:11796-00000-20
Specifications
About this item
Virtua™ Sport protective eyewear are sleek and unisex. These are features with a Corded Earplug Control System (CCS).
- Large wraparound lens keeps eyes more fully protected and provides a wide viewing area
- Contoured temples fit comfortably and snugly without pressure points
- Impact-resistant polycarbonate lens absorbs 99.9% UV rays
Virtua™ Sport Protective Eyewear lends an athletic-inspired image in an economy line of safety glasses that offers both protection and affordability.
Eye protection is important to safeguard the user's eyes and maintain healthy vision. According to the Bureau of Labor Statistics, nearly three out of every five workers who suffered eye injuries were not wearing eye protection at the time of the accident. Wearing the correct safety eyewear for the task and for the unique needs of the worker is important. All 3M™ protective eyewear combines safety features with comfort enhancements.
These safety glasses are recommended for use in chipping, chiseling, drilling, grinding, machining, masonry, pouring/casting, power fastening, riveting, sanding, and sawing applications. Industries in which these safety glasses are commonly used include aviation, chemicals, construction, food and beverage, general manufacturing, mining, oil and gas, pharmaceuticals, primary metals, and transportation.