BBL™ Dextrose Tryptone Agar, BD Biosciences
Catalog # 90004-020
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Specifications
- Description:BBL™ Dextrose Tryptone Agar
- Size:500 g
- Cat. no.:90004-020
- Supplier No.:280100
Specifications
About this item
Dextrose Tryptone Agar is used for cultivating thermophilic "flat-sour" microorganisms associated with food spoilage.
- Peptones provide carbon and nitrogen sources
- Dextrose is the carbohydrate source
- Bromcresol purple is the pH indicator
- Agar is the solidifying agent
Dextrose Tryptone Agar can also be used to isolate other food from spoilage bacteria; mesophilic aerobic spore formers in the genera Bacillus and Sporolactobacillus and thermophilic flat sour spore formers such as B. stearothermophilus.