Order Entry
Mexico
ContactUsLinkComponent
Difco™ Biotin Assay Medium, BD
Catalog # 90003-264
CAS Number:  
Difco™ Biotin Assay Medium, BD
Catalog # 90003-264
Supplier Number:  241910
CAS Number:  
Specifications
  • Description:
    Difco™ Biotin Assay Medium
  • Size:
    100 g
  • Cat. no.:
    90003-264
  • Supplier No.:
    241910
Specifications
About this item
Biotin Assay Medium is used for determining biotin concentration by the microbiological assay technique.
  • Biotin-free dehydrated medium
  • Addition of biotin gives a growth response
  • Dehydrated appearance: Light beige, homogeneous
Biotin Assay Medium may be used for both turbidimetric and titrimetric analysis. The most effective assay range, using Biotin Assay Medium, has been found to be between 0.1 ng and 1 ng biotin.