Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Difco™ B12 Assay Medium
- Size:100 g
- Cat. no.:90003-384
- Supplier No.:245710
Specifications
About this item
B12 Assay Medium is used for determining vitamin B12 concentration by the microbiological assay technique.
- Dehydrated appearance: Very light to light beige, homogeneous
- 4.25% single strength solution
- Solution is light amber, clear, may have a slight precipitate
B12 Assay Medium is used in the microbiological assay of vitamin B12 according to the procedures of the Vitamin B12 Activity Assay in the USP1 and the Cobalamin (Vitamin B12 Activity) Assay in the Official Methods of Analysis of AOAC International (AOAC).