Order Entry
Mexico
Orders LinkContactUsLinkComponent
Difco™ Pseudomonas Isolation Agar, BD

Difco™ Pseudomonas Isolation Agar, BD

Difco™ Pseudomonas Isolation Agar, 500 g
Catalog # 90004-394
Supplier #:  292710
Difco™ Pseudomonas Isolation Agar, BD
Catalog # 90004-394
CAS Number:  
Difco™ Pseudomonas Isolation Agar, BD

Difco™ Pseudomonas Isolation Agar, 500 g

CAS Number:  
Price
$6,600.00
Log in for availability
Each (500g)
Product Details & Documents

Pseudomonas Isolation Agar is used with added glycerol in isolating Pseudomonas and differentiating Pseudomonas aeruginosa from other pseudomonads based on pigment formation.

  • Peptone provides a source of carbon and nitrogen
  • Magnesium chloride and potassium sulfate promote production of pyocyanin
  • Irgasan selectively inhibits contaminant bacteria
  • Agar is the solidifying agent

Glycerol serves as an energy source and also helps to promote pyocyanin production.

Documents

Specifications

Specifications

  • Description:
    Difco™ Pseudomonas Isolation Agar
  • Size:
    500 g

Specifications