Plasmodium falciparum, Gametocytes , Thin Smear Slide
Catalog # 470181-952
Supplier: Avantor
Specifications
- Description:Plasmodium falciparum, Gametocytes , Thin Smear
- Cat. no.:470181-952
- Stain:Giemsa
- Supplier No.:WARD470181-952
- Orientation:Smear
Specifications
About this item
Can Cause Malaria in Humans
- Identifying features clearly distinguishable
- Rigorous quality control standards
Microscope slide showing the parasitic protozoan Plasmodium falciparum in human blood.
To learn more about this organism, please visit the CDC's article about the stages of Plasmodium falciparum in blood: https://www.cdc.gov/dpdx/resources/pdf/benchAids/malaria/Pfalciparum_benchaidV2.pdf