Order Entry
Mexico
Orders LinkContactUsLinkComponent
Medullated Nerve (Mammal)
Medullated Nerve (Mammal), Cross Section and Longitudinal Section, Harris, Phloxine and Saffron
  470177-600
 :   Avantor
Medullated Nerve (Mammal)
  470177-600
 :  Avantor
Medullated Nerve (Mammal)
Medullated Nerve (Mammal)
Medullated Nerve (Mammal), Cross Section and Longitudinal Section, Harris, Phloxine and Saffron
$193.60
Each (1UN)
Investigate Nerve Tissue.
  • All identifying features are clearly distinguishable
  • Rigorous quality control standards
Harris, phloxine, and safranin, cross section and longitudinal section. Peripheral nerve bundles showing axis cylinders, medullary sheaths, Schwann nuclei, epi- and perineurium.
  • Description:
    Medullated Nerve (Mammal)
  • Stain:
    Harris, Phloxine and Saffron
  • Orientation:
    Cross Section and Longitudinal Section