Thermometer Clamp
Thermometer Clamp
470019-488
: THCL01
A must have for any lab.
- Adjustable 360° rotating jaw
- Attaches to rods up to 13mm (1/2") in diameter
Use this thermometer clamp to safely hold a thermometer in place during experiments.
- Description:Thermometer Clamp
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



