BBL™ Salmonella Shigella Agar (SS Agar), BD
Catálogo # 90000-240
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Specifications
- Description:BBL™ Salmonella Shigella Agar (SS Agar)
- Size:500 g
- Cat. no.:90000-240
- Supplier No.:211597
Specifications
About this item
Salmonella Shigella Agar is a moderately selective and differential medium for the isolation, cultivation, and differentiation of Gram-negative enteric microorganisms isolated from both clinical and nonclinical specimens.
- Moderately selective medium
- Contains bile salts, brilliant green, and citrates
Salmonella Shigella Agar is designated as a moderately selective medium based upon the degree of inhibition of Gram-positive microorganisms which it inhibits due to its content of bile salts, brilliant green, and citrates.