Order Entry
Mexico
ContactUsLinkComponent
Rappaport-Vassiliadis R10 Broth, BD Diagnostics
Rappaport-Vassiliadis R10 Broth, BD Diagnostics
Catalog # 90001-010
CAS Number:  
Rappaport-Vassiliadis R10 Broth, BD Diagnostics
Catalog # 90001-010
Supplier Number:  218581
CAS Number:  
Specifications
  • Description:
    Rappaport-Vassiliadis R10 Broth
  • Size:
    500 g
  • Cat. no.:
    90001-010
  • Supplier No.:
    218581
Specifications
About this item
Rappaport-Vassiliadis R10 Broth is a liquid medium for selectively enriching Salmonella from meat and dairy products, feces, and polluted water.
  • Tryptone provides carbon and nitrogen for general growth
  • Malachite green is inhibitory to organisms other than Salmonella
  • Low pH helps select for Salmonella species
Selective media recommended by the European Chocolate Manufacturer's Association and the AOAC for rapid detection of motile Salmonella.