N,N-Dimethylacetamide ≥99.5% BDH HiPerSolv CHROMANORM for HPLC
Catalog # BDH83636.400
Proveedor: VWR International
Número CAS:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Especificaciones
- Pack type:Glass bottle, amber, poly-coated
- Size:4 L
- Cat. no.:BDH83636.400
Especificaciones
Resultados de prueba de especificación
- Assay (by GC-FID):Min 99.5%
- Residue after evaporation:Max 5 ppm
- Water (by KF coulometry):Max 0.03%
- Transmittance at 270 nm:Min 10 T(%)
- Transmittance at 280 nm:Min 50 T(%)
- Transmittance at 290 nm:Min 70 T(%)
- Transmittance at 310 nm:Min 90 T(%)
- Transmittance at 320 nm:Min 93 T(%)
- Transmittance at 360 nm:Min 97 T(%)
- Transmittance at 400 nm:Min 97 T(%)
- Optical absorbance at 270 nm:Max 1.00 AU
- Optical absorbance at 280 nm:Max 0.30 AU
- Optical absorbance at 290 nm:Max 0.15 AU
- Optical absorbance at 320 AU:Max 0.05 AU
- Optical absorbance at 310 nm:Max 0.03 AU
- Optical absorbance at 360 nm:Max 0.01 AU
- Optical absorbance at 400 nm:Max 0.01 AU
Resultados de prueba de especificación
Acerca de este artículo
:
:87.12 g/mol
Punto de ebullición:166.1 °C (1013 hPa)
Punto de ebullición:166.1 °C (1013 hPa)
Punto de fusión:–20 °C
Punto de fusión:-20 °C
Punto de inflamación:70 °C
UN:2810
ADR:6.1,III
Número CAS:127-19-5
Fórmula:CH₃CON(CH₃)₂
Número MDL:MFCD00008686
Punto de inflamación:70 °C